5-Chloro-2-(pivaloylamino)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31076-1G 31076-25G 31076-500MG 31076-5G
76930-532EA 379.81 USD
76930-532 76930-534 76930-536 76930-538
5-Chloro-2-(pivaloylamino)phenylboronic acid pinacol ester ≥97%
5-Chloro-2-(pivaloylamino)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BClNO₃
MW: 337.649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380464
CAS Number: 1409999-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR