5-(Methothio)thiophene-2-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31026-1G 31026-25G 31026-5G
76930-518EA 109.28 USD
76930-518 76930-520 76930-522
5-(Methothio)thiophene-2-boronic acid pinacol ester ≥97%
5-(Methothio)thiophene-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₂S₂
Storage Temperature: Ambient
MDL Number: MFCD16660065
CAS Number: 1172999-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR