2-(4-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-methoxyphenylboronic acid pinacol ester

31020-1G 31020-25G 31020-5G
76930-506EA 62.6 USD
76930-506 76930-508 76930-510
2-(4-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176538
CAS Number: 627525-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR