5-Chloro-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(5-Chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Chloro-2-fluorobenzeneboronic acid pinacol ester

31016-25G 31016-5G
76930-494EA 275.84 USD
76930-494 76930-496
5-Chloro-2-fluorophenylboronic acid pinacol ester ≥97%
5-Chloro-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16036133
CAS Number: 1190129-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR