5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

30975-1G 30975-25G 30975-500MG 30975-5G
76930-480EA 133.68 USD
76930-480 76930-482 76930-484 76930-486
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester ≥97%
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester
Formula: C₁₂H₁₇BClNO₃
MW: 269.53 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063054
CAS Number: 1083168-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR