5-Bromo-3-Methyl-2-(Methylsulfonyl)Pyridine ≥97%

Supplier: AOBChem USA

Warning

30873-1G 30873-25G 30873-500MG 30873-5G
76930-456EA 260.99 USD
76930-456 76930-458 76930-460 76930-462
5-Bromo-3-Methyl-2-(Methylsulfonyl)Pyridine ≥97%
5-Bromo-3-Methyl-2-(Methylsulfonyl)Pyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂S
MW: 250.113 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28130266
CAS Number: 1445651-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR