5-Chloro-2,6-dimethoxy-3-pyridinecarboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

30824-1G 30824-25G 30824-500MG 30824-5G
76930-440EA 350.1 USD
76930-440 76930-442 76930-444 76930-446
5-Chloro-2,6-dimethoxy-3-pyridinecarboxaldehyde ≥97%
5-Chloro-2,6-dimethoxy-3-pyridinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088216
CAS Number: 1879026-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR