4-Bromo-3-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30807-1G 30807-25G 30807-500MG 30807-5G
76930-424EA 290.69 USD
76930-424 76930-426 76930-428 76930-430
4-Bromo-3-hydroxyphenylboronic acid ≥97%
4-Bromo-3-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BBrO₃
Storage Temperature: Ambient
MDL Number: MFCD22491266
CAS Number: 1701448-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR