4-Chloro-2,3-difluoro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30790-1G 30790-25G 30790-500MG 30790-5G
76930-394EA 379.81 USD
76930-394 76930-396 76930-398 76930-400
4-Chloro-2,3-difluoro-5-methoxybenzaldehyde ≥97%
4-Chloro-2,3-difluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088207
CAS Number: 1879026-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR