4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-3-(trifluoromethyl)phenylboronic acid pinacol ester

30781-1G 30781-25G 30781-500MG 30781-5G
76930-378EA 173.99 USD
76930-378 76930-380 76930-382 76930-384
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoic acid ≥97% research grade
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoic acid
Formula: C₁₄H₁₆BF₃O₄
MW: 316.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28356850
CAS Number: 2121513-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR