5-Bromo-2-(trifluoromethoxy)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30701-1G 30701-25G 30701-500MG 30701-5G
76930-356EA 133.68 USD
76930-356 76930-358 76930-360 76930-362
5-Bromo-2-(trifluoromethoxy)benzyl alcohol ≥97%
5-Bromo-2-(trifluoromethoxy)benzyl alcohol
Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03701031
CAS Number: 685126-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR