4-Methoxy-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

30623-1G 30623-25G 30623-5G
76930-344EA 100.79 USD
76930-344 76930-346 76930-348
4-Methoxy-3-(trifluoromethyl)phenol ≥97%
4-Methoxy-3-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09832303
CAS Number: 53903-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR