5-Bromo-1-fluoro-3-iodo-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

30453-1G 30453-25G 30453-500MG 30453-5G
76930-316EA 250.38 USD
76930-316 76930-318 76930-320 76930-322
5-Bromo-1-fluoro-3-iodo-2-methoxybenzene ≥97%
5-Bromo-1-fluoro-3-iodo-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Ambient
CAS Number: 1228093-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR