4-Bromo-3,5-dichlorophenyl boronic acid ≥97%

Supplier: AOBChem USA

Warning

30361-1G 30361-25G 30361-500MG 30361-5G
76930-264EA 275.84 USD
76930-264 76930-266 76930-268 76930-270
4-Bromo-3,5-dichlorophenyl boronic acid ≥97%
4-Bromo-3,5-dichlorophenyl boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrCl₂O₂
MW: 269.716 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088158
CAS Number: 2121511-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR