4-Chloro-2,3-dimethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30356-1G 30356-500MG 30356-5G
76930-250EA 528.33 USD
76930-250 76930-252 76930-254
4-Chloro-2,3-dimethylphenylboronic acid pinacol ester ≥97%
4-Chloro-2,3-dimethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BClO₂
MW: 266.571 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088155
CAS Number: 2121514-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR