5-Chloro-2-ethoxy-nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-ethoxynicotinic acid

30328-25G 30328-5G
76930-240EA 350.1 USD
76930-240 76930-242
5-Chloro-2-ethoxy-nicotinic acid ≥97%
5-Chloro-2-ethoxy-nicotinic acid
Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07403147
CAS Number: 68359-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR