5,6-Dichloro-2-iodo-3-pyridinol ≥95%

Supplier: AOBChem USA

Warning

30158-1G 30158-25G 30158-500MG 30158-5G
76930-172EA 263.11 USD
76930-172 76930-174 76930-176 76930-178
5,6-Dichloro-2-iodo-3-pyridinol ≥95%
5,6-Dichloro-2-iodo-3-pyridinol

Precaución: for Research Use Only

Formula: C₅H₂Cl₂INO
MW: 289.886 g/mol
Storage Temperature: Ambient
CAS Number: 188057-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR