1-Iodo-2-methyl-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Iodo-3-methylbenzotrifluoride

29983-1G 29983-25G 29983-5G
76930-130EA 114.58 USD
76930-130 76930-132 76930-134
1-Iodo-2-methyl-4-(trifluoromethyl)benzene ≥97%
1-Iodo-2-methyl-4-(trifluoromethyl)benzene
Formula: C₈H₆F₃I
MW: 286.0329 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570417
CAS Number: 54978-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR