4-Bromo-3-chloro-2-methylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

29915-1G 29915-25G 29915-5G
76930-116EA 94.43 USD
76930-116 76930-118 76930-120
4-Bromo-3-chloro-2-methylphenylboronic acid pinacol ester ≥95%
4-Bromo-3-chloro-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrClO₂
Storage Temperature: Ambient
MDL Number: MFCD29036611
CAS Number: 1799612-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR