4-Bromo-3-chloro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29912-1G 29912-500MG 29912-5G
76930-102EA 468.92 USD
76930-102 76930-104 76930-106
4-Bromo-3-chloro-2-methylbenzoic acid ≥97%
4-Bromo-3-chloro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19686176
CAS Number: 1349708-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR