5-Bromo-2-ethoxypyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

29597-1G 29597-25G 29597-5G
76930-066EA 94.43 USD
76930-066 76930-068 76930-070
5-Bromo-2-ethoxypyridine-4-carboxylic acid ≥97%
5-Bromo-2-ethoxypyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17019252
CAS Number: 1379305-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR