4-Isopropoxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29571-25G 29571-5G
76930-058EA 524.09 USD
76930-058 76930-060
4-Isopropoxy-3-methoxybenzaldehyde ≥97%
4-Isopropoxy-3-methoxybenzaldehyde
Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00837588
CAS Number: 2538-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR