4-Hydroxy-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

29496-25G 29496-5G
76930-038EA 289.63 USD
76930-038 76930-040
4-Hydroxy-2-(trifluoromethyl)benzoic acid ≥97%
4-Hydroxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01091007
CAS Number: 320-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR