4-Chloro-1-ethyl-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

29340-1G 29340-25G 29340-500MG 29340-5G
76930-026EA 144.29 USD
76930-026 76930-028 76930-030 76930-032
4-Chloro-1-ethyl-2-nitrobenzene ≥97%
4-Chloro-1-ethyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Refrigerator
CAS Number: 2001-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR