5-Methoxymethyl-thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Methoxymethyl)thiophene-2-carboxylic acid

28796-1G 28796-25G 28796-500MG 28796-5G
76929-904EA 221.73 USD
76929-904 76929-906 76929-908 76929-910
5-Methoxymethyl-thiophene-2-carboxylic acid ≥97%
5-Methoxymethyl-thiophene-2-carboxylic acid
Formula: C₇H₈O₃S
MW: 172.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02735563
CAS Number: 61855-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR