4-Hydroxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28710-25G 28710-5G
76929-892EA 225.98 USD
76929-892 76929-894
4-Hydroxy-2-methylbenzaldehyde ≥97%
4-Hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.1479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037443
CAS Number: 41438-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR