4-Cyclohexylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28614-1G 28614-25G 28614-500MG 28614-5G
76929-870EA 260.99 USD
76929-870 76929-872 76929-874 76929-876
4-Cyclohexylbenzaldehyde ≥97%
4-Cyclohexylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₆O
Storage Temperature: Refrigerator
CAS Number: 27634-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR