4-Cyclobutylphenol ≥95%

Supplier: AOBChem USA

Warning

28586-1G 28586-25G 28586-500MG 28586-5G
76929-860EA 296 USD
76929-860 76929-862 76929-864 76929-866
4-Cyclobutylphenol ≥95%
4-Cyclobutylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
Storage Temperature: Refrigerator
MDL Number: MFCD21099815
CAS Number: 10292-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR