4-Cyclobutylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

28579-1G 28579-25G 28579-500MG 28579-5G
76929-846EA 320.4 USD
76929-846 76929-848 76929-850 76929-852
4-Cyclobutylbenzaldehyde ≥95%
4-Cyclobutylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD19440003
CAS Number: 875306-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR