4-Bromo-2-ethylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

28568-1G 28568-25G 28568-500MG 28568-5G
76929-832EA 379.81 USD
76929-832 76929-834 76929-836 76929-838
4-Bromo-2-ethylbenzonitrile ≥95%
4-Bromo-2-ethylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD18826345
CAS Number: 744200-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR