4-Hydroxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28526-25G 28526-5G
76929-820EA 212.18 USD
76929-820 76929-822
4-Hydroxy-2-methoxybenzaldehyde ≥97%
4-Hydroxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051964
CAS Number: 18278-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR