5-Bromo-2,3-dichlorotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1,2-dichloro-3-methylbenzene

28486-1G 28486-25G 28486-500MG 28486-5G
76929-804EA 155.96 USD
76929-804 76929-806 76929-808 76929-810
5-Bromo-2,3-dichlorotoluene ≥97%
5-Bromo-2,3-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.925 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878183
CAS Number: 204930-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR