5-Bromo-2-chloro-3-(pyrrolidine-1-carbonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

28452-1G 28452-25G 28452-500MG 28452-5G
76929-790EA 236.59 USD
76929-790 76929-792 76929-794 76929-796
5-Bromo-2-chloro-3-(pyrrolidine-1-carbonyl)pyridine ≥97%
5-Bromo-2-chloro-3-(pyrrolidine-1-carbonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂O
MW: 289.556 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16159731
CAS Number: 1249608-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR