5-Ethyl-2-thiophenecarboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

28375-25G 28375-5G
76929-778EA 279.02 USD
76929-778 76929-780
5-Ethyl-2-thiophenecarboxaldehyde ≥97%
5-Ethyl-2-thiophenecarboxaldehyde
Formula: C₇H₈OS
Storage Temperature: Ambient
MDL Number: MFCD00143457
CAS Number: 36880-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR