5-Butylthiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

28345-1G 28345-25G 28345-5G
76929-766EA 114.58 USD
76929-766 76929-768 76929-770
5-Butylthiophene-2-carbaldehyde ≥97%
5-Butylthiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₂OS
MW: 168.256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03872257
CAS Number: 98954-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR