4-Fluoro-1-benzofuran-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-benzofuran-2-carboxylic acid

28256-1G 28256-25G 28256-500MG 28256-5G
76929-742EA 379.81 USD
76929-742 76929-744 76929-746 76929-748
4-Fluoro-1-benzofuran-2-carboxylic acid ≥97%
4-Fluoro-1-benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅FO₃
MW: 180.133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23708314
CAS Number: 1408060-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR