4-Fluorobenzofuran-7-carbaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

28204-1G 28204-25G 28204-500MG 28204-5G
76929-732EA 171.87 USD
76929-732 76929-734 76929-736 76929-738
4-Fluorobenzofuran-7-carbaldehyde ≥97% research grade
4-Fluorobenzofuran-7-carbaldehyde
Formula: C₉H₅FO₂
MW: 164.133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11052336
CAS Number: 555155-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR