3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Hydroxy-2-methoxyphenylboronic acid pinacol ester

28044-1G 28044-25G 28044-500MG 28044-5G
76929-688EA 245.07 USD
76929-688 76929-690 76929-692 76929-694
3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formula: C₁₃H₁₉BO₄
MW: 250.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994337
CAS Number: 507462-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR