4-Chloro-2-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

27994-25G 27994-5G
76929-676EA 559.1 USD
76929-676 76929-678
4-Chloro-2-(trifluoromethyl)benzyl alcohol ≥97%
4-Chloro-2-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04115839
CAS Number: 773872-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR