4-Fluoro-1-naphthaldehyde ≥95%

Supplier: AOBChem USA

Warning

27927-25G 27927-5G
76929-648EA 595.17 USD
76929-648 76929-650
4-Fluoro-1-naphthaldehyde ≥95%
4-Fluoro-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₇FO
MW: 174.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02261765
CAS Number: 172033-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR