4-Cyclopropyl-3-mehtoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27860-1G 27860-500MG 27860-5G
76929-638EA 498.63 USD
76929-638 76929-640 76929-642
4-Cyclopropyl-3-mehtoxyphenylboronic acid ≥97%
4-Cyclopropyl-3-mehtoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
MW: 192.019 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21099609
CAS Number: 1840935-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR