4-Bromo-3-(trifluoromethoxy)phenol ≥97%
Supplier: AOBChem USA
|
Warning
|
27703-25G
27703-5G
76929-606EA
354.35
USD
76929-606
76929-608
4-Bromo-3-(trifluoromethoxy)phenol ≥97%
4-Bromo-3-(trifluoromethoxy)phenol
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD06660164 CAS Number: 886499-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...