4-Chloro-2,6-dimethylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

27639-1G 27639-25G 27639-500MG 27639-5G
76929-594EA 231.28 USD
76929-594 76929-596 76929-598 76929-600
4-Chloro-2,6-dimethylbenzyl alcohol ≥97%
4-Chloro-2,6-dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD28142605
CAS Number: 332179-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR