4-Bromo-2-fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

27537-1G 27537-25G 27537-500MG 27537-5G
76929-540EA 205.82 USD
76929-540 76929-542 76929-544 76929-546
4-Bromo-2-fluoro-3-iodobenzaldehyde ≥97%
4-Bromo-2-fluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrFIO
MW: 328.905 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142600
CAS Number: 1809158-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR