5-Butylthiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

27388-1G 27388-25G 27388-5G
76929-486EA 62.6 USD
76929-486 76929-488 76929-490
5-Butylthiophene-2-carboxylic acid ≥97%
5-Butylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD20639640
CAS Number: 63068-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR