5-Bromo-6-chloropicolinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-6-chloropyridine-2-carboxylic acid

27344-1G 27344-25G 27344-500MG 27344-5G
76929-472EA 221.73 USD
76929-472 76929-474 76929-476 76929-478
5-Bromo-6-chloropicolinic acid ≥97%
5-Bromo-6-chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09910220
CAS Number: 959958-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR