2-(5-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Hexylthiopheneboronic acid pinacol ester

27213-1G 27213-25G 27213-5G
76929-446EA 96.55 USD
76929-446 76929-448 76929-450
2-(5-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(5-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₆H₂₇BO₂S
MW: 294.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10567052
CAS Number: 917985-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR