4-Fluoro-2-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

27169-1G 27169-25G 27169-5G
76929-436EA 66.84 USD
76929-436 76929-438 76929-440
4-Fluoro-2-isopropoxybenzaldehyde ≥97%
4-Fluoro-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015440
CAS Number: 1289162-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR