5-Chloro-2-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27009-1G 27009-25G 27009-5G
76929-384EA 84.88 USD
76929-384 76929-386 76929-388
5-Chloro-2-ethoxybenzoic acid ≥97%
5-Chloro-2-ethoxybenzoic acid
Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD02729233
CAS Number: 62871-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR