5-(Benzyloxy)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

26999-1G 26999-25G 26999-500MG 26999-5G
76929-364EA 296 USD
76929-364 76929-366 76929-368 76929-370
5-(Benzyloxy)nicotinic acid ≥97%
5-(Benzyloxy)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11845573
CAS Number: 263270-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR